Gene Transcripts

Gene Symbol
Sat2
Entrez Gene ID
69215
Gene Name
spermidine/spermine N1-acetyl transferase 2
Gene Aliases
2610016A03Rik, SSAT-2, SSAT2
Chromosome Location
Chr. 11: 69621993 - 69624034 on Build GRCm38
UniGene ID
Mm.340410
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026991.2 NP_081267.1 6 604
GenBank mRNA AK011406.1 - 7 523
BC061227.1 - 6 604 30312881

View Full Product Details