Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000247.2 | NP_000238.1 | 3 | 641 | |
NM_001177519.2 | NP_001170990.1 | 3 | 641 | ||
NM_001289152.1 | NP_001276081.1 | 3 | 1505 | ||
NM_001289153.1 | NP_001276082.1 | 3 | 1485 | ||
NM_001289154.1 | NP_001276083.1 | 3 | 1313 | ||
GenBank mRNA | AB506764.1 | - | 3 | 575 | |
AK094237.1 | - | 3 | 1313 | ||
AK225393.1 | - | 3 | 623 | ||
AK299135.1 | - | 3 | 613 | ||
AK310028.1 | - | 3 | 584 | ||
AK311667.1 | - | 1 | 1504 | ||
AY204547.1 | - | 3 | 575 | ||
AY750850.1 | - | 3 | 575 | ||
BC016929.1 | - | 3 | 629 | 4446575 | |
BM552351.1 | - | 3 | 569 | ||
BX342160.2 | - | 3 | 599 | ||
L14848.1 | - | 3 | 614 |