Gene Transcripts

Gene Symbol
Ubl7
Entrez Gene ID
69459
Gene Name
ubiquitin-like 7 (bone marrow stromal cell-derived)
Gene Aliases
2300004C15Rik
Chromosome Location
Chr. 9: 57910980 - 57929970 on Build GRCm38
UniGene ID
Mm.41735
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122873.1 NP_001116345.1 3, 4 399
NM_027086.3 NP_081362.2 3, 4 347
GenBank mRNA AK009025.1 - 3, 4 348
AK154496.1 - 3, 4 357
BC016456.1 - 2, 3 333 4912715
BC094047.1 - 2, 3 336 5253101
CX203425.1 - 3, 4 377

View Full Product Details