Gene Transcripts

Gene Symbol
Klk12
Entrez Gene ID
69511
Gene Name
kallikrein related-peptidase 12
Gene Aliases
2310008B01Rik, AI324852, KLK-L5
Chromosome Location
Chr. 7: 43768664 - 43774704 on Build GRCm38
UniGene ID
Mm.20163
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027097.1 NP_081373.1 2 160
GenBank mRNA AK009217.1 - 1 23
BC147442.1 - 2 160 9056084
BC147463.1 - 2 160 9056105

View Full Product Details