Gene Transcripts

Gene Symbol
Pla2g12b
Entrez Gene ID
69836
Gene Name
phospholipase A2, group XIIB
Gene Aliases
2010002E04Rik, Fksg71, Pla2g13, hlb218
Chromosome Location
Chr. 10: 59403420 - 59421976 on Build GRCm38
UniGene ID
Mm.30268
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023530.2 NP_076019.2 2 349
GenBank mRNA AF339738.1 - 2 371
AK008035.1 - 2 375
AY358033.1 - 2 378
BC021592.1 - 2 349 5133141

View Full Product Details