Gene Transcripts

Gene Symbol
Smc4
Entrez Gene ID
70099
Gene Name
structural maintenance of chromosomes 4
Gene Aliases
2500002A22Rik, C79747, SMC-4, Smc4l1
Chromosome Location
Chr. 3: 69004972 - 69034623 on Build GRCm38
UniGene ID
Mm.206841
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133786.3 NP_598547.1 4 550
GenBank mRNA AJ534940.2 - 3 426
AK088350.1 - 4 533
AK088846.1 - 4 516
AK133817.1 - 4 540
AK152804.1 - 4 577
BC061481.1 - 4 525 6512832
BC062939.1 - 4 550 6841276

View Full Product Details