Gene Transcripts

Gene Symbol
Poc1a
Entrez Gene ID
70235
Gene Name
POC1 centriolar protein homolog A (Chlamydomonas)
Gene Aliases
2510040D07Rik, Wdr51a
Chromosome Location
Chr. 9: 106281024 - 106349891 on Build GRCm38
UniGene ID
Mm.23054
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027354.2 NP_081630.2 6, 7 1049
GenBank mRNA AK011064.1 - 6, 7 727
AK154163.1 - 6, 7 799
BC034901.1 - 6, 7 709 5255069

View Full Product Details