Gene Transcripts

Gene Symbol
Mospd1
Entrez Gene ID
70380
Gene Name
motile sperm domain containing 1
Gene Aliases
1810018L05Rik
Chromosome Location
Chr. X: 53344594 - 53370562 on Build GRCm38
UniGene ID
Mm.491083
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290514.1 NP_001277443.1 3 407
NM_027409.5 NP_081685.1 3 407
GenBank mRNA AK007533.1 - 3 340
AK088275.1 - 3 344
AK139795.1 - 3 351
BC018329.1 - 3 308 3962906
BC085100.1 - 3 315 6391017
BC100442.1 - 3 295 30501889
CJ046213.1 - 3 408

View Full Product Details