Gene Transcripts

Gene Symbol
Ggnbp1
Entrez Gene ID
70772
Gene Name
gametogenetin binding protein 1
Gene Aliases
0610031G08Rik, mdf
Chromosome Location
Chr. 17: 27018035 - 27036378 on Build GRCm38
UniGene ID
Mm.440437
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001251881.1 NP_001238810.1 3 367
NM_027544.2 NP_081820.2 2 796
GenBank mRNA AA388242.1 - 3 338
AB250379.1 - 2 679
AK077129.1 - 2 679
AY141192.1 - 2 796
BC049764.1 - 2 657 6774330
CF585840.1 - 3 379

View Full Product Details