Gene Transcripts
- Gene Symbol
- Mier1
- Entrez Gene ID
- 71148
- Gene Name
- mesoderm induction early response 1 homolog (Xenopus laevis
- Gene Aliases
- 4933425I22Rik, 5830411K19Rik, er1
- Chromosome Location
- Chr. 4: 103114390 - 103165754 on Build GRCm38
- UniGene ID
- Mm.288721
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039081.2 |
NP_001034170.1
|
11, 12 |
1187 |
|
NM_001286221.1 |
NP_001273150.1
|
13, 14 |
1318 |
|
NM_001286222.1 |
NP_001273151.1
|
13, 14 |
1372 |
|
NM_001286223.1 |
NP_001273152.1
|
11, 12 |
1168 |
|
NM_027696.3 |
NP_081972.2
|
13, 14 |
1375 |
|
GenBank mRNA |
AK016915.1 |
- |
5, 6 |
617 |
|
AK129405.1 |
- |
13, 14 |
1295 |
|
AK136467.1 |
- |
1, 2 |
31 |
|
AY769435.1 |
- |
12, 13 |
1167 |
|
AY769436.1 |
- |
11, 12 |
1090 |
|
AY769437.1 |
- |
11, 12 |
1140 |
|
AY769438.1 |
- |
13, 14 |
1240 |
|
BC108273.1 |
- |
11, 12 |
1154 |
30080243 |
BC130227.1 |
- |
11, 12 |
1139 |
40141863 |
CN701986.1 |
- |
5, 6 |
541 |
|
View Full Product Details