Gene Transcripts

Gene Symbol
MRC1
Entrez Gene ID
4360
Gene Name
mannose receptor, C type 1
Gene Aliases
CD206, CLEC13D, CLEC13DL, MMR, MRC1L1, bA541I19.1, hMR
Chromosome Location
Chr. 10: 17809343 - 17911162 on Build GRCh38
UniGene ID
Hs.743903
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002438.3 NP_002429.1 2 485
GenBank mRNA AK308132.1 - 2 486
AK308167.1 - 2 481
BC142642.1 - 2 466
BC146838.1 - 2 408 9056765
DC296466.1 - 2 485
DQ663787.1 - 2 362
J05550.1 - 2 465
X55635.1 - 2 438

View Full Product Details