Gene Transcripts

Gene Symbol
Cdc5l
Entrez Gene ID
71702
Gene Name
cell division cycle 5-like (S. pombe)
Gene Aliases
1200002I02Rik, AA408004, PCDC5RP
Chromosome Location
Chr. 17: 45391887 - 45433707 on Build GRCm38
UniGene ID
Mm.28270
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152810.2 NP_690023.1 13 2023
GenBank mRNA AK004547.2 - 13 2053
AK146892.1 - 13 1843
AK150562.1 - 13 1840
AK150861.1 - 13 1840
AK172950.1 - 13 1843
BC003893.1 - 14 1958 3593535
BC031480.1 - 13 2023 3498853

View Full Product Details