Gene Transcripts
- Gene Symbol
- R3hcc1
- Entrez Gene ID
- 71843
- Gene Name
- R3H domain and coiled-coil containing 1
- Gene Aliases
- 1700020M16Rik, AA407796
- Chromosome Location
- Chr. 14: 69697304 - 69714809 on Build GRCm38
- UniGene ID
- Mm.27738
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001146012.2 |
NP_001139484.2
|
5, 6 |
1270 |
|
NM_001301651.1 |
NP_001288580.1
|
5, 6 |
1222 |
|
NM_001301652.1 |
NP_001288581.1
|
4, 5 |
1503 |
|
GenBank mRNA |
AK006177.1 |
- |
5, 6 |
1222 |
|
AK032854.1 |
- |
5, 6 |
1240 |
|
AK156843.1 |
- |
5, 6 |
1240 |
|
AK206441.1 |
- |
|
100 |
|
AK208982.1 |
- |
1, 2 |
100 |
|
BC052536.1 |
- |
4, 5 |
1419 |
|
BC059860.1 |
- |
4, 5 |
1503 |
|
BQ747890.1 |
- |
1, 2 |
134 |
|
View Full Product Details