Gene Transcripts

Gene Symbol
Dpep3
Entrez Gene ID
71854
Gene Name
dipeptidase 3
Gene Aliases
1700018F16Rik, AI507039, MBD-3, MLZ-136
Chromosome Location
Chr. 8: 105973520 - 105979419 on Build GRCm38
UniGene ID
Mm.173395
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027960.2 NP_082236.1 8 1164
GenBank mRNA AF488553.1 - 8 1045
AK006085.1 - 8 1165
BC051148.1 - 8 1174 6704315

View Full Product Details