Gene Transcripts
- Gene Symbol
- Suds3
- Entrez Gene ID
- 71954
- Gene Name
- suppressor of defective silencing 3 homolog (S. cerevisiae)
- Gene Aliases
- 2400003N08Rik, 2410008L21Rik, AU067672, mSds3
- Chromosome Location
- Chr. 5: 117091678 - 117115993 on Build GRCm38
- UniGene ID
- Mm.34456
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001122666.2 |
NP_001116138.1
|
6, 7 |
698 |
|
NM_178622.5 |
NP_848737.3
|
6, 7 |
698 |
|
GenBank mRNA |
AF469109.1 |
- |
6, 7 |
638 |
|
AK010274.1 |
- |
6, 7 |
602 |
|
AK043652.1 |
- |
6, 7 |
641 |
|
AK045475.1 |
- |
6, 7 |
652 |
|
AK138947.1 |
- |
6, 7 |
649 |
|
AK150633.1 |
- |
6, 7 |
636 |
|
AK188755.1 |
- |
1, 2 |
118 |
|
BC036562.1 |
- |
1, 2 |
71 |
|
BC040088.1 |
- |
4, 5 |
311 |
|
BC055764.1 |
- |
6, 7 |
651 |
6402735 |
BC062176.1 |
- |
6, 7 |
565 |
6514171 |
View Full Product Details