Gene Transcripts

Gene Symbol
Ugt2a3
Entrez Gene ID
72094
Gene Name
UDP glucuronosyltransferase 2 family, polypeptide A3
Gene Aliases
2010321J07Rik
Chromosome Location
Chr. 5: 87324972 - 87337195 on Build GRCm38
UniGene ID
Mm.482739
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028094.3 NP_082370.2 6 1509
GenBank mRNA AK008601.1 - 6 1491
AK050327.1 - 6 1510
BC025795.1 - 6 1485 5133743

View Full Product Details