Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028339.1 | NP_082615.1 | 4 | 418 | |
GenBank mRNA | AK013150.1 | - | 4 | 418 | |
AK030429.1 | - | 4 | 436 | ||
AK050567.1 | - | 4 | 417 | ||
AK137059.1 | - | 4 | 415 | ||
AK150511.1 | - | 4 | 425 | ||
AK153027.1 | - | 4 | 421 | ||
AK170468.1 | - | 4 | 436 | ||
BC017603.1 | - | 4 | 418 | 4503129 | |
BC021533.1 | - | 4 | 439 | 5354337 |