Gene Transcripts

Gene Symbol
Osbp2
Entrez Gene ID
74309
Gene Name
oxysterol binding protein 2
Gene Aliases
1700095P05Rik, C630001G20Rik, Gm244, ORP-4, OSBPL1
Chromosome Location
Chr. 11: 3703731 - 3863907 on Build GRCm38
UniGene ID
Mm.61022
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152818.2 NP_690031.2 11 2176
GenBank mRNA AK007088.1 - 7 757
AK044598.1 - 8 965
BC031794.1 - 9 1139 4501119
BC034567.1 - 1 16
BC051185.1 - 9 1136
BC058356.1 - 10 1319 5696357
BC058602.1 - 9 1139 6467653

View Full Product Details