Gene Transcripts

Gene Symbol
Clec16a
Entrez Gene ID
74374
Gene Name
C-type lectin domain family 16, member A
Gene Aliases
4932416N17Rik, curt, mKIAA0350
Chromosome Location
Chr. 16: 10545339 - 10744878 on Build GRCm38
UniGene ID
Mm.159176
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204229.1 NP_001191158.1 4 606
NM_177562.5 NP_808230.2 4 606
GenBank mRNA AK089327.1 - 4 537
AK122255.1 - 4 558
AK155314.1 - 4 548
AK156927.1 - 4 537
AK170754.1 - 4 537
AK171134.1 - 4 537
BC138907.1 - 4 509 8861929
BC138909.1 - 4 509 8861931

View Full Product Details