Gene Transcripts

Gene Symbol
Fam53a
Entrez Gene ID
74504
Gene Name
family with sequence similarity 53, member A
Gene Aliases
2410018C17Rik, 5430419M09Rik, Dntnp
Chromosome Location
Chr. 5: 33600351 - 33629647 on Build GRCm38
UniGene ID
Mm.294695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178390.3 NP_848477.2 4, 5 709
GenBank mRNA AF396665.1 - 2, 3 131
AK010537.1 - 4, 5 646
AK017321.1 - 3, 4 1052
AK163326.1 - 4, 5 680
AK215806.1 - 78
AK220276.1 - 3, 4 345
BC016596.1 - 2, 3 134 3257498
BC028656.1 - 4, 5 429 1244982

View Full Product Details