Gene Transcripts

Gene Symbol
Fam63a
Entrez Gene ID
75007
Gene Name
family with sequence similarity 63, member A
Gene Aliases
1810005H09Rik, 4930504E06Rik, AA562754, AA960384, AW561904, cI-40, mKIAA1390, ni
Chromosome Location
Chr. 3: 95281590 - 95296164 on Build GRCm38
UniGene ID
Mm.279287
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133858.4 NP_598619.2 4 1556
NM_199475.1 NP_955769.1 4 1556
GenBank mRNA AB075603.1 - 3 567
AK036838.1 - 4 1548
AK087859.1 - 4 1369
AK162551.1 - 4 1276
AK169673.1 - 4 1556
AK208613.1 - 3 189
BC039762.1 - 3 374
BC051048.1 - 3 439
BC057901.1 - 2 85
BC094388.1 - 4 1373 5708548
BC132301.1 - 3 801 40130578
BC144749.1 - 3 801 9053297

View Full Product Details