Gene Transcripts

Gene Symbol
Fabp12
Entrez Gene ID
75497
Gene Name
fatty acid binding protein 12
Gene Aliases
1700008G05Rik
Chromosome Location
Chr. 3: 10244209 - 10301526 on Build GRCm38
UniGene ID
Mm.47052
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029310.1 NP_083586.1 4 465
GenBank mRNA AK005765.1 - 3 311
BC030910.1 - 4 465 4505673
EU733649.1 - 3 441

View Full Product Details