Gene Transcripts
- Gene Symbol
- NDUFB8
- Entrez Gene ID
- 4714
- Gene Name
- NADH:ubiquinone oxidoreductase subunit B8
- Gene Aliases
- ASHI, CI-ASHI
- Chromosome Location
- Chr. 10: 100523729 - 100529923 on Build GRCh38
- UniGene ID
- Hs.523215
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001284368.1 |
NP_001271297.1
|
4, 5 |
458 |
|
NM_005004.3 |
NP_004995.1
|
4, 5 |
549 |
|
GenBank mRNA |
AF044958.1 |
- |
4, 5 |
499 |
|
AF115968.1 |
- |
4, 5 |
505 |
|
AK289579.1 |
- |
4, 5 |
490 |
|
AL080056.1 |
- |
4, 5 |
509 |
|
BC000466.2 |
- |
4, 5 |
490 |
2822781 |
BC019276.1 |
- |
4, 5 |
490 |
2822781 |
BM008944.1 |
- |
4, 5 |
446 |
|
CF127298.1 |
- |
4, 5 |
488 |
|
CR533490.1 |
- |
4, 5 |
477 |
|
View Full Product Details