Gene Transcripts

Gene Symbol
DRG1
Entrez Gene ID
4733
Gene Name
developmentally regulated GTP binding protein 1
Gene Aliases
NEDD3
Chromosome Location
Chr. 22: 31399553 - 31434186 on Build GRCh38
UniGene ID
Hs.115242
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004147.3 NP_004138.1 4, 5 554
GenBank mRNA AF078103.1 - 4, 5 484
AJ005940.1 - 4, 5 488
AK098576.1 - 4, 5 430
AK315659.1 - 4, 5 512
BC019285.1 - 4, 5 493 2823263
BC020803.1 - 4, 5 488 4250998
BT007237.1 - 4, 5 423
CR456488.1 - 4, 5 465
CR542059.1 - 4, 5 423

View Full Product Details