Gene Transcripts
- Gene Symbol
- NFKB2
- Entrez Gene ID
- 4791
- Gene Name
- nuclear factor kappa B subunit 2
- Gene Aliases
- CVID10, H2TF1, LYT-10, LYT10, NF-kB2, p100, p49/p100, p52
- Chromosome Location
- Chr. 10: 102394110 - 102402529 on Build GRCh38
- UniGene ID
- Hs.73090
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001077494.3 |
NP_001070962.1
|
19, 20 |
2493 |
|
NM_001261403.2 |
NP_001248332.1
|
18, 19 |
2588 |
|
NM_001288724.1 |
NP_001275653.1
|
19, 20 |
2787 |
|
NM_001322934.1 |
NP_001309863.1
|
19, 20 |
2497 |
|
NM_001322935.1 |
NP_001309864.1
|
18, 19 |
2661 |
|
NM_002502.5 |
NP_002493.3
|
19, 20 |
2497 |
|
XM_011539830.2 |
XP_011538132.1
|
14, 15 |
1851 |
|
XM_011539831.2 |
XP_011538133.1
|
14, 15 |
1848 |
|
XM_017016278.1 |
XP_016871767.1
|
19, 20 |
2782 |
|
GenBank mRNA |
AK098415.1 |
- |
9, 10 |
1370 |
|
AK292654.1 |
- |
18, 19 |
2441 |
|
BC002844.2 |
- |
19, 20 |
2324 |
3634575 |
BT009769.1 |
- |
18, 19 |
2224 |
|
S76638.1 |
- |
19, 20 |
2474 |
|
View Full Product Details