Gene Transcripts

Gene Symbol
Ermn
Entrez Gene ID
77767
Gene Name
ermin, ERM-like protein
Gene Aliases
A330104H05Rik, AI854460, ermin, mKIAA1189
Chromosome Location
Chr. 2: 58045115 - 58052752 on Build GRCm38
UniGene ID
Mm.91712
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029972.3 NP_084248.2 1 176
GenBank mRNA AK020733.1 - 1 288
AK078089.1 - 1 266
AK136820.1 - 1 266
AK137305.1 - 1 268
AK220372.1 - 1 295
BC089515.1 - 1 177 6589784
BC090670.1 - 1 208 30941185
DQ334270.1 - 1 154

View Full Product Details