Gene Transcripts

Gene Symbol
Tchp
Entrez Gene ID
77832
Gene Name
trichoplein, keratin filament binding
Gene Aliases
A930031F18Rik, AV279199
Chromosome Location
Chr. 5: 114707765 - 114722327 on Build GRCm38
UniGene ID
Mm.239698
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029992.2 NP_084268.2 2 319
GenBank mRNA AK020912.1 - 2 282
AK083027.1 - 2 235
AK159947.1 - 2 280
AK216033.1 - 1 37
BC043048.1 - 2 224 5704036
BU512279.1 - 2 319

View Full Product Details