Gene Transcripts

Gene Symbol
Yipf6
Entrez Gene ID
77929
Gene Name
Yip1 domain family, member 6
Gene Aliases
A430107J06Rik
Chromosome Location
Chr. X: 98936321 - 98949021 on Build GRCm38
UniGene ID
Mm.212290
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207633.2 NP_997516.1 3 671
GenBank mRNA AK020780.1 - 3 245
AK030473.1 - 3 673
AK033612.1 - 1 250
AK045453.1 - 3 383
AK053562.1 - 3 672
AK079683.1 - 3 283
AK143152.1 - 3 241
AK160251.1 - 4 299
BC048712.1 - 3 306 6328657

View Full Product Details