Gene Transcripts
- Gene Symbol
- Mccc2
- Entrez Gene ID
- 78038
- Gene Name
- methylcrotonoyl-Coenzyme A carboxylase 2 (beta)
- Gene Aliases
- 4930552N12Rik, MCCB
- Chromosome Location
- Chr. 13: 99948532 - 100015639 on Build GRCm38
- UniGene ID
- Mm.137327
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030026.2 |
NP_084302.1
|
14, 15 |
1505 |
|
GenBank mRNA |
AK132265.1 |
- |
14, 15 |
1485 |
|
AK143233.1 |
- |
14, 15 |
1506 |
|
AK145564.1 |
- |
14, 15 |
1457 |
|
AK146844.1 |
- |
14, 15 |
1465 |
|
AK146865.1 |
- |
14, 15 |
1455 |
|
BC033050.1 |
- |
1, 2 |
167 |
|
BC040098.1 |
- |
6, 7 |
517 |
|
BC051500.1 |
- |
2, 3 |
324 |
|
BC058987.1 |
- |
6, 7 |
519 |
|
BC141193.1 |
- |
14, 15 |
1471 |
9055805 |
BC141194.1 |
- |
14, 15 |
1471 |
9055806 |
View Full Product Details