Gene Transcripts

Gene Symbol
Spaca7
Entrez Gene ID
78634
Gene Name
sperm acrosome associated 7
Gene Aliases
1700094C09Rik
Chromosome Location
Chr. 8: 12572997 - 12600744 on Build GRCm38
UniGene ID
Mm.116871
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024279.2 NP_077241.1 1, 2 154
GenBank mRNA AK018926.1 - 1, 2 155
BC048522.1 - 1, 2 154 6741699

View Full Product Details