Gene Transcripts

Gene Symbol
Popdc3
Entrez Gene ID
78977
Gene Name
popeye domain containing 3
Gene Aliases
AA682104, Pop3
Chromosome Location
Chr. 10: 45289305 - 45318450 on Build GRCm38
UniGene ID
Mm.24748
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024286.1 NP_077248.1 2 908
GenBank mRNA AK039691.1 - 2 812
AK052325.1 - 2 908
BC114993.1 - 2 731 40056724

View Full Product Details