Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030694.1 | NP_109619.1 | 1 | 176 | |
GenBank mRNA | AK141392.1 | - | 1 | 203 | |
AY082486.1 | - | 1 | 198 | ||
BC002102.1 | - | 1 | 176 | 3485116 | |
BC002160.1 | - | 1 | 181 | 3485614 | |
BC084679.1 | - | 1 | 176 | 5374182 |