Gene Transcripts

Gene Symbol
Cnnm2
Entrez Gene ID
94219
Gene Name
cyclin M2
Gene Aliases
AU015877, AW048635, Acdp2, Clp2
Chromosome Location
Chr. 19: 46760381 - 46878580 on Build GRCm38
UniGene ID
Mm.306903
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102471.1 NP_001095941.1 2, 3 1930
NM_033569.3 NP_291047.2 2, 3 1930
GenBank mRNA AF216961.1 - 2, 3 1242
AK032006.1 - 2, 3 1889
AK053711.1 - 2, 3 1948
AK159616.1 - 2, 3 1936
BC125033.1 - 2, 3 1242
GQ225745.1 - 2, 3 1229

View Full Product Details