Gene Transcripts

Gene Symbol
Olah
Entrez Gene ID
99035
Gene Name
oleoyl-ACP hydrolase
Gene Aliases
AI607300, E230009B14Rik, Thedc1
Chromosome Location
Chr. 2: 3341982 - 3397238 on Build GRCm38
UniGene ID
Mm.13808
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145921.1 NP_666033.1 4 490
GenBank mRNA AK087571.1 - 4 510
AK136031.1 - 5 746
BC025001.1 - 4 490 5252879

View Full Product Details