Gene Transcripts

Gene Symbol
ROR1
Entrez Gene ID
4919
Gene Name
receptor tyrosine kinase like orphan receptor 1
Gene Aliases
NTRKR1, dJ537F10.1
Chromosome Location
Chr. 1: 63774019 - 64181498 on Build GRCh38
UniGene ID
Hs.128753
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083592.1 NP_001077061.1 7 1538
NM_005012.3 NP_005003.2 7 1538
XM_011541526.1 XP_011539828.1 7 1137
XM_017001376.1 XP_016856865.1 6 1438
XM_017001377.1 XP_016856866.1 9 4081
GenBank mRNA AK308088.1 - 7 1544
AK309189.1 - 4 780
BC080541.1 - 7 1538
BC128386.1 - 8 1148
M97675.1 - 7 1514

View Full Product Details