Gene Transcripts

Gene Symbol
Mtus1
Entrez Gene ID
102103
Gene Name
mitochondrial tumor suppressor 1
Gene Aliases
AI481402, ATBP135, Atip1, B430010I23Rik, B430305I03Rik, C85752, Cctsg1-440, MD44, MTSG1, mKIAA1288
Chromosome Location
Chr. 8: 40990912 - 41133983 on Build GRCm38
UniGene ID
Mm.149438
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005863.2 NP_001005863.1 9, 10 3443
NM_001005864.3 NP_001005864.1 7, 8 1049
NM_001005865.3 NP_001005865.2 5, 6 1062
NM_001286413.1 NP_001273342.1 9, 10 3325
GenBank mRNA AF173380.1 - 5, 6 901
AF493235.1 - 5, 6 966
AK030510.1 - 5, 6 1026
AK031693.1 - 5, 6 996
AK035576.1 - 3, 4 430
AY246699.1 - 5, 6 724
AY626781.1 - 5, 6 1059
AY626782.1 - 7, 8 1047
AY626783.1 - 8, 9 3265
BC041777.1 - 8, 9 2108 4240274
BC042206.1 - 8, 9 2108 4240274
BC043321.1 - 7, 8 1013
BC089009.1 - 9, 10 3325 30543240

View Full Product Details