Gene Transcripts

Gene Symbol
Pwp1
Entrez Gene ID
103136
Gene Name
PWP1 homolog (S. cerevisiae)
Gene Aliases
2310058A11Rik, 2610205J09Rik, AI098170, AL022879, AV083409, AW550690
Chromosome Location
Chr. 10: 85871831 - 85889103 on Build GRCm38
UniGene ID
Mm.18834
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133993.3 NP_598754.2 8 898
GenBank mRNA AK009972.1 - 8 865
AK011891.1 - 8 846
AK012649.1 - 2 141
AK075805.1 - 8 840
AK082158.1 - 8 840
AK149699.1 - 8 843
BC003199.1 - 8 867 3586496
BC023137.1 - 8 848 3986908

View Full Product Details