Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001287188.1 | NP_001274117.1 | 8 | 1152 | |
NM_001287189.1 | NP_001274118.1 | 8 | 1144 | ||
NM_001287190.1 | NP_001274119.1 | 8 | 990 | ||
NM_002539.2 | NP_002530.1 | 8 | 1252 | ||
GenBank mRNA | AK292352.1 | - | 8 | 1125 | |
AK301956.1 | - | 8 | 925 | ||
AK312766.1 | - | 6 | 692 | ||
BC025296.1 | - | 8 | 992 | 5088190 | |
M16650.2 | - | 7 | 778 | ||
M20372.1 | - | 4 | 359 | ||
X55362.1 | - | 8 | 993 |