Gene Transcripts
- Gene Symbol
- Ypel1
- Entrez Gene ID
- 106369
- Gene Name
- yippee-like 1 (Drosophila)
- Gene Aliases
- 0610009L05Rik, 1700016N17Rik, 1700019O22Rik, 4921520K19Rik, 4930511F14Rik, AV118478, Dgl1, Ppil2, mdgl-1
- Chromosome Location
- Chr. 16: 17070138 - 17086736 on Build GRCm38
- UniGene ID
- Mm.237941
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001291047.1 |
NP_001277976.1
|
5, 6 |
786 |
|
NM_001291053.1 |
NP_001277982.1
|
3, 4 |
380 |
|
NM_023249.6 |
NP_075738.1
|
4, 5 |
660 |
|
GenBank mRNA |
AB098740.1 |
- |
3, 4 |
272 |
|
AF190624.1 |
- |
4, 5 |
576 |
|
AK005955.1 |
- |
4, 5 |
496 |
|
AK006137.1 |
- |
3, 4 |
278 |
|
AK006505.1 |
- |
4, 5 |
471 |
|
AK006858.1 |
- |
4, 5 |
548 |
|
AK011452.1 |
- |
4, 5 |
591 |
|
AK014931.1 |
- |
4, 5 |
486 |
|
AK015752.1 |
- |
3, 4 |
497 |
|
AK018910.1 |
- |
4, 5 |
471 |
|
AK019819.1 |
- |
4, 5 |
491 |
|
AK076063.1 |
- |
4, 5 |
603 |
|
AK133401.1 |
- |
4, 5 |
574 |
|
AK139453.1 |
- |
4, 5 |
568 |
|
BB610926.1 |
- |
|
699 |
|
BC049737.1 |
- |
3, 4 |
290 |
6771362 |
BQ573264.1 |
- |
3, 4 |
346 |
|
View Full Product Details