Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001291047.1 | NP_001277976.1 | 3 | 568 | |
NM_023249.6 | NP_075738.1 | 2 | 442 | ||
GenBank mRNA | AB098740.1 | - | 1 | 54 | |
AF190624.1 | - | 2 | 358 | ||
AK005955.1 | - | 2 | 278 | ||
AK006049.1 | - | 2 | 278 | ||
AK006505.1 | - | 2 | 253 | ||
AK006858.1 | - | 2 | 330 | ||
AK014931.1 | - | 2 | 268 | ||
AK015752.1 | - | 1 | 279 | ||
AK018910.1 | - | 2 | 253 | ||
AK019819.1 | - | 2 | 273 | ||
AK076063.1 | - | 2 | 385 | ||
AK133401.1 | - | 2 | 356 | ||
AK139453.1 | - | 2 | 350 | ||
AK163365.1 | - | 3 | 509 | ||
BB610926.1 | - | 3 | 481 | ||
BB649357.1 | - | 2 | 442 |