Gene Transcripts

Gene Symbol
ACYP2
Entrez Gene ID
98
Gene Name
acylphosphatase 2
Gene Aliases
ACYM, ACYP
Chromosome Location
Chr. 2: 53970838 - 54305300 on Build GRCh38
UniGene ID
Hs.516173
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320586.1 NP_001307515.1 7 707
NM_001320587.1 NP_001307516.1 6 614
NM_001320588.1 NP_001307517.1 4 408
NM_001320589.1 NP_001307518.1 5 729
NM_138448.3 NP_612457.1 4 659
XM_017005411.1 XP_016860900.1 8 1053
XM_017005412.1 XP_016860901.1 9 1123
XM_017005413.1 XP_016860902.1 9 1094
GenBank mRNA BC012290.1 - 4 428
BF671963.1 - 4 407
BG741807.1 - 6 608
BU543943.1 - 7 707
BU857820.1 - 5 587
X84195.1 - 4 408

View Full Product Details