Gene Transcripts

Gene Symbol
P2RX5
Entrez Gene ID
5026
Gene Name
purinergic receptor P2X 5
Gene Aliases
LRH-1, P2X5, P2X5R
Chromosome Location
Chr. 17: 3673227 - 3696404 on Build GRCh38
UniGene ID
Hs.731607
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204519.1 NP_001191448.1 1 452
NM_001204520.1 NP_001191449.1 1 452
NM_002561.3 NP_002552.2 1 452
NM_175080.2 NP_778255.1 1 452
GenBank mRNA AF016709.1 - 1 89
AF070573.1 - 1 80
AK290889.1 - 1 452
AK307959.1 - 1 283
BC039015.1 - 1 193 5773706
BM477082.1 - 338
DQ234349.1 - 1 53
U49395.1 - 1 105
U49396.1 - 1 105

Gene Symbol
P2RX5-TAX1BP3
Entrez Gene ID
100533970
Gene Name
P2RX5-TAX1BP3 readthrough (NMD candidate)
Gene Aliases
Chromosome Location
Chr. 17: 3662893 - 3696404 on Build GRCh38
UniGene ID
Hs.731607
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037928.1 1 452
GenBank mRNA AB209623.1 - 1 242

View Full Product Details