Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001167914.1 | NP_001161386.1 | 4 | 396 | |
NM_029705.3 | NP_083981.2 | 4 | 396 | ||
GenBank mRNA | AF537188.1 | - | 4 | 310 | |
AK008675.1 | - | 4 | 355 | ||
AK030842.1 | - | 4 | 336 | ||
BC049743.1 | - | 4 | 396 | ||
BC087880.1 | - | 4 | 391 | 6771498 |