Gene Transcripts

Gene Symbol
Prlr
Entrez Gene ID
19116
Gene Name
prolactin receptor
Gene Aliases
AI987712, Pr-1, Pr-3, Prlr-rs1
Chromosome Location
Chr. 15: 10177238 - 10349190 on Build GRCm38
UniGene ID
Mm.10516
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011169.5 NP_035299.4 10 5537
GenBank mRNA AK087727.1 - 1 401
BC096586.1 - 1 175
DV053280.1 - 1 595

View Full Product Details