Gene Transcripts

Gene Symbol
DPP9
Entrez Gene ID
91039
Gene Name
dipeptidyl peptidase 9
Gene Aliases
DP9, DPLP9, DPRP-2, DPRP2
Chromosome Location
Chr. 19: 4675226 - 4723843 on Build GRCh38
UniGene ID
Hs.515081
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139159.4 NP_631898.3 9 1246
XM_005259673.2 XP_005259730.1 10 1355
XM_011528405.1 XP_011526707.1 11 1407
XM_011528406.1 XP_011526708.1 12 1517
XM_011528408.1 XP_011526710.1 9 1192
XM_011528410.1 XP_011526712.1 10 1407
XM_017027453.1 XP_016882942.1 11 1997
XM_017027454.1 XP_016882943.1 12 2107
GenBank mRNA AF452102.2 - 9 1266
AF542510.2 - 9 1225
AK122645.1 - 3 296
AK122654.1 - 9 1249
AK131100.1 - 6 815
AK131499.1 - 8 759
AL834376.1 - 3 277
AY172660.1 - 6 911
AY374518.1 - 6 901
BC037948.2 - 9 1183 4577856
DQ417928.1 - 7 988

View Full Product Details