Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001359.1 | NP_001350.1 | 9 | 1058 | |
XM_005250808.4 | XP_005250865.1 | 8 | 746 | ||
XM_011516880.2 | XP_011515182.1 | 10 | 1053 | ||
XM_017013147.1 | XP_016868636.1 | 11 | 1713 | ||
XM_017013148.1 | XP_016868637.1 | 11 | 1453 | ||
GenBank mRNA | AK300069.1 | - | 11 | 1573 | |
BC105080.1 | - | 9 | 1005 | 8144083 | |
BC105082.1 | - | 9 | 1005 | 8144085 | |
BC143669.1 | - | 7 | 702 | ||
L26050.1 | - | 9 | 927 |