Gene Transcripts

Gene Symbol
LEPR
Entrez Gene ID
3953
Gene Name
leptin receptor
Gene Aliases
CD295, LEP-R, LEPRD, OB-R, OBR
Chromosome Location
Chr. 1: 65420652 - 65637493 on Build GRCh38
UniGene ID
Hs.23581
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002303.5 NP_002294.2 20 3544
GenBank mRNA BC131779.1 - 6 1297
U43168.1 - 20 3552
U66497.1 - 19 3485

View Full Product Details