Gene Transcripts

Gene Symbol
SPR
Entrez Gene ID
6697
Gene Name
sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase)
Gene Aliases
SDR38C1
Chromosome Location
Chr. 2: 72887383 - 72892160 on Build GRCh38
UniGene ID
Hs.301540
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003124.4 NP_003115.1 3 1229
GenBank mRNA AK222942.1 - 3 1217
BC017310.2 - 3 1193 5089862

View Full Product Details