Gene Transcripts
- Gene Symbol
- EHHADH
- Entrez Gene ID
- 1962
- Gene Name
- enoyl-CoA, hydratase/3-hydroxyacyl CoA dehydrogenase
- Gene Aliases
- ECHD, FRTS3, L-PBE, LBFP, LBP, PBFE
- Chromosome Location
- Chr. 3: 185190624 - 185282886 on Build GRCh38
- UniGene ID
- Hs.429879
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
L07077.1 |
- |
2 |
118 |
|
View Full Product Details