Gene Transcripts

Gene Symbol
MKNK1
Entrez Gene ID
8569
Gene Name
MAP kinase interacting serine/threonine kinase 1
Gene Aliases
MNK1
Chromosome Location
Chr. 1: 46557407 - 46607140 on Build GRCh38
UniGene ID
Hs.371594
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135553.2 NP_001129025.1 13 1374
NM_003684.5 NP_003675.2 14 1497
NM_198973.3 NP_945324.1 12 1288
NR_024174.2 11 1256
NR_024176.2 13 1401
XM_006711000.1 XP_006711063.1 13 1327
XM_006711001.2 XP_006711064.1 14 1577
XM_006711002.2 XP_006711065.1 13 6802
XM_006711003.1 XP_006711066.1 14 1388
XM_006711006.3 XP_006711069.1 12 4117
XM_006711007.1 XP_006711070.1 12 1219
XM_006711009.3 XP_006711072.1 12 6704
XM_006711013.1 XP_006711076.1 12 1252
XM_006711014.1 XP_006711077.1 12 1243
XM_006711015.1 XP_006711078.1 13 1314
XM_017002652.1 XP_016858141.1 13 7050
XM_017002653.1 XP_016858142.1 14 1675
XM_017002654.1 XP_016858143.1 14 4527
XM_017002655.1 XP_016858144.1 14 7148
XM_017002656.1 XP_016858145.1 14 6947
XM_017002657.1 XP_016858146.1 13 6849
XM_017002658.1 XP_016858147.1 12 7006
XM_017002659.1 XP_016858148.1 12 1302
XM_017002660.1 XP_016858149.1 12 6758
XM_017002661.1 XP_016858150.1 13 1523
XM_017002662.1 XP_016858151.1 13 1344
XM_017002663.1 XP_016858152.1 13 7104
XM_017002664.1 XP_016858153.1 12 6728
XM_017002665.1 XP_016858154.1 11 1175
XM_017002666.1 XP_016858155.1 12 4159
XM_017002667.1 XP_016858156.1 10 1735
GenBank mRNA AB000409.1 - 13 1317
AB451300.1 - 12 1130
AB451432.1 - 12 1130
AK096423.1 - 11 963
AK290456.1 - 12 1250
AK298839.1 - 11 1230
AY355461.1 - 11 1086
BC002755.2 - 14 1417 3629765
BX538193.1 - 11 1124

View Full Product Details